missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ribophorin II Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA555266
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: SSGYYDFLVE VEGDNRYIAN TVELRVKIST EVGITNVDLS TVDKDQSIAP KTTRVTYPAK AKGTFIADSH QNFALFFQLV DVNTGAELTP HQTFVRLHNQ KTGQEVVFVA EPDNKNVYKF ELDTSERKIE FD Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
RNP2 a type I integral membrane protein found only in the rough endoplasmic reticulum. The encoded protein is part of an N-oligosaccharyl transferase complex that links high mannose oligosaccharides to asparagine residues found in the Asn-X-Ser/Thr consensus motif of nascent polypeptide chains. This protein is similar in sequence to the yeast oligosaccharyl transferase subunit SWP1.
Especificaciones
| Ribophorin II | |
| Polyclonal | |
| Unconjugated | |
| RPN2 | |
| 1300012C06Rik; AV261018; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 63 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 2; hypothetical protein LOC534231; oligosaccharyltransferase complex subunit (non-catalytic); RIBIIR; ribophorin 2; ribophorin II; Ribophorin-2; RPN2; Rpn-2; RPNII; RPN-II; SWP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6185 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P04844 | |
| RPN2 | |
| Recombinant protein corresponding to Human RPN2. Recombinant protein control fragment (Product #RP-89456). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido