missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RI-2, MedChemExpress
Description
RI-2 is a reversible RAD51 inhibitor, with an IC50 of 44.17 μM, and specifically inhibits homologous recombination repair in human cells.
Specifications
Specifications
| Nombre del producto químico o material | RI-2 |
| CAS | 1417162-36-7 |
| Color | Naranja |
| Notas de grado de pureza | Research |
| Fórmula molecular | C21H18Cl2N2O4 |
| Cantidad | 100 mg |
| Información de solubilidad | DMSO : 130 mg/mL (300.04 mM; Need ultrasonic) |
| SMILES | O=C(C(C1=CC=C(OC)C=C1)=C2N3CCOCC3)N(C4=CC=C(Cl)C(Cl)=C4)C2=O |
| Peso molecular (g/mol) | 433.28 |
| Formula Weight (peso de la fórmula) | 433.28 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?