missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RhoA Monoclonal Antibody (1B8-1C7)

Código de producto. 15347364
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15347364 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15347364 Proveedor Invitrogen™ N.º de proveedor MA1134

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

By Western blot, MA1-134 detects recombinant RhoA, but not recombinant RhoB or recombinant RhoC. By Western blot on mouse and rat kidney whole tissue lysates, MA1-134 detects a band of ~22kD corresponding to the expected molecular weight of RhoA. A nonspecific band at ~55kD was also detected in both kidney lysates.

RhoA regulates a signal transduction pathway linking plasma membrane receptors to the assembly of focal adhesions and actin stress fibers. It is involved in a microtubule-dependent signal that is required for the myosin contractile ring formation during cell cycle cytokinesis, and plays an essential role in cleavage furrow formation. RhoA is required for the apical junction formation of keratinocyte cell-cell adhesion, and serves as a target for the yopT cysteine peptidase from Yersinia pestis, vector of the plague, and Yersinia pseudotuberculosis, which causes gastrointestinal disorders. RhoA stimulates PKN2 kinase activity and may be an activator of PLCE1. It is activated by ARHGEF2, which promotes the exchange of GDP for GTP. It is essential for the SPATA13-mediated regulation of cell migration and adhesion assembly and disassembly. The MEMO1-RHOA-DIAPH1 signaling pathway plays an important role in ERBB2-dependent stabilization of microtubules at the cell cortex. It controls the localization of APC and CLASP2 to the cell membrane, via the regulation of GSK3B activity. In turn, membrane-bound APC allows the localization of the MACF1 to the cell membrane, which is required for microtubule capture and stabilization.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno RhoA
Aplicaciones Western Blot
Clasificación Monoclonal
Clon 1B8-1C7
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1mg/mL BSA and 0.05% sodium azide
génica RHOA
N.º de referencia del gen P61586, P61589, Q9QUI0
Alias de gen Aplysia ras-related homolog 12; Aplysia RAS-related homolog 6; aplysia ras-related homolog A; aplysia ras-related homolog A1; aplysia ras-related homolog A2; ARH12; ARH6; ARH9; ARHA; Arha1; Arha2; ARHB; ARHC; h12; h6; H9; MGC72339; MST081; MSTP081; oncogene RHO H12; oncogene RHO H6; oncogene RHO H9; plysia ras-related homolog A2; Ras family member A; ras homolog A1; ras homolog A2; ras homolog family member A; ras homolog family member B; ras homolog family member C; ras homolog gene family, member A; ras homolog gene family, member A1; ras homolog gene family, member A2; ras homolog gene family, member B; ras homolog gene family, member C; RAS-related homolog 9; Rho; Rho cDNA clone 12; Rho cDNA clone 6; rho cDNA clone 9; Rho family GTPase; RHO12; Rhoa; RHOB; RHOC; rhoC GTPase; RHOH12; RHOH6; RHOH9; rho-related GTP-binding protein RhoB; Rho-related GTP-binding protein RhoC; small GTP binding protein RhoA; small GTP binding protein RhoC; transforming protein RhoA
Símbolos de los genes RHOA
Especie del huésped Mouse
Inmunógeno Full-length recombinant His-tagged human RhoA.
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 117273, 11848, 387
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.