Learn More
Invitrogen™ Rheumatoid Factor Monoclonal Antibody (RF2-17-109.2), Invitrogen™
Código de producto. p-7245247
Change view
Click to view available options
Cantidad:
1 mg
100 μg
500 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
500 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
17880286 100 μg 100 microgramos
11528882 500 μg 500 microgramos
17850356 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 17880286 Proveedor Invitrogen™ N.º de proveedor MA545978
Mouse Monoclonal Antibody
This antibody reacts with 30% of IgM-RF from unrelated individuals. It does not recognize IgM, IgA, or IgG proteins lacking RF nor with F(ab’)2 fragments.
Antibodies directed against the Fc fragment of immunoglobulin G (IgG) are called rheumatoid factors (RFs). They are heterogenous and are usually IgM antibodies. RFs are used as a marker in individuals with suspected rheumatoid arthritis (RA) or other autoimmune conditions. The normal reference range for RF is less than 15 IU/mL.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Rheumatoid Factor
Aplicaciones ELISA
Clasificación Monoclonal
Clon RF2-17-109.2
Concentración 1.8 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative; pH 7.4
Alias de gen RF
Especie del huésped Mouse
Inmunógeno Purified proteins from patients with IgM cryoglobulins.
Método de purificación Protein G
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b
Vis mere Vis mindre
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.