missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS17 Monoclonal Antibody (OTI8G4), TrueMAB™, OriGene
Descripción
RGS17 Monoclonal antibody specifically detects RGS17 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Especificaciones
Especificaciones
| Antígeno | RGS17 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI8G4 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | PYGM |
| N.º de referencia del gen | P09812, P11217, Q9WUB3 |
| Alias de gen | 6430507P11Rik; hRGS17; regulator of G-protein signaling 17; regulator of G-protein signaling Z2; regulator of Gz-selective protein signaling 2; Rgs17; RGS-17; Rgsz2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?