Learn More
Invitrogen™ RGMA Recombinant Rabbit Monoclonal Antibody (24GB1320)
GREENER_CHOICE
Código de producto. 30276196
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
30276196 100 μL 100 microlitros
30276599 20 μL 20 microlitros
2 options
Código de producto. 30276196
Marca: Invitrogen™ MA557221
Rabbit Recombinant Monoclonal Antibody
RGMA Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Member of the repulsive guidance molecule (RGM) family that performs several functions in the developing and adult nervous system. Regulates cephalic neural tube closure, inhibits neurite outgrowth and cortical neuron branching, and the formation of mature synapses. Binding to its receptor NEO1/neogenin induces activation of RHOA-ROCK1/Rho-kinase signaling pathway through UNC5B-ARHGEF12/LARG-PTK2/FAK1 cascade, leading to collapse of the neuronal growth cone and neurite outgrowth inhibition. Furthermore, RGMA binding to NEO1/neogenin leads to HRAS inactivation by influencing HRAS-PTK2/FAK1-AKT1 pathway. It also functions as a bone morphogenetic protein (BMP) coreceptor that may signal through SMAD1, SMAD5, and SMAD8.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno RGMA
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 24GB1320
Concentración 0.67 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica RGMA
N.º de referencia del gen Q6PCX7, Q96B86
Alias de gen BC059072; C230063O06; repulsive guidance molecule; repulsive guidance molecule A; repulsive guidance molecule family member a; RGM; RGM domain family member A; RGM domain family, member A; RGMA
Símbolos de los genes RGMA
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human RGMA (aa 401-435).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 244058, 308739, 56963
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG