Learn More
STEMCELL Technologies ReproRNA™-OKSGM Kit
Click to view available options
Cantidad:
1 Kit
Tamaño de la unidad:
1 set
Descripción
- ReproRNA™-OKSGM is a single-stranded RNA replicon vector that contains five reprogramming factors: OCT4, KLF-4, SOX2, GLIS1, and c-MYC, as well as a puromycin-resistance gene (Figure 1)
- This RNA vector reprograms somatic cells, such as fibroblasts, into induced pluripotent stem (iPS) cells with high efficiency and only requires a single transfection step (Figure 2)
- When used together with ReproTeSR™ reprogramming medium, the generation of iPS cell colonies can be achieved under feeder-free conditions with superior colony morphology and similar reprogramming efficiency to feeder-based systems (Figures 3 - 4)
- ReproRNA™-derived iPS cell colonies also express markers of undifferentiated cells and retain a normal karyotype (Figures 5-6)
- Subsequently, iPS cells generated with ReproRNA™-OKSGM can be maintained in TeSR™ maintenance media (mTeSR™1, TeSR™2, or TeSR™-E8™) and further differentiated into cells of all three germ layers (Figure 7)
Especificaciones
Especificaciones
| Tipo de célula | Pluripotent Stem Cells |
| Para utilizar con (aplicación) | Cell Culture; Reprogramming |
| Cantidad | 1 Kit |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.