Learn More
Grant Instruments™ Tapas transparentes de policarbonato
Product Code. 11486417
Click to view available options
Para utilizar con (equipo):
Baño de agua modelo SAP12, JBN12, JBA12 y SBB Aqua 12 Plus SUB Aqua Plus
Depósito de 2 l
Depósito de 5 l
Depósitos de 26 l
Unit Size:
1 PCS
cada uno
This item is not returnable. View return policy
Product Code. 11486417
Brand: Grant Instruments™ AQL5
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Dirige la condensación lejos de los recipientes sumergidos, evita la contaminación, reduce la evaporación y ahorra energía.
  • Diseño de policarbonato
  • La tapa transparente de dos vertientes permite que la acumulación de condensación gotee de los recipientes sumergidos.
  • Para su uso con el baño de agua sin agitación Sub Aqua Pro
TRUSTED_SUSTAINABILITY
Specifications
Descripción En dos vertientes
Para utilizar con (equipo) Depósito de 5 l
Material Policarbonato
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.