Learn More
BOLA Replacement Inner Parts
Description
- Consisting of a v-ring (ETFE), a tapered ring and a sealing ring (both PTFE) as well as an o-ring (FKM) for tubing diameters under 3 mm (not exposed to the medium).
- Very good chemical resistance, the medium is only exposed to PTFE.
Specifications
Specifications
| Certificaciones/conformidad | FDA |
| Diámetro (métrico) | 3 mm |
| Esterilizables en autoclave | Autoclavable |
| Tamaño de rosca | GL 25 |
| Color | White |
| Flexibility | Rigid |