Learn More
Thermo Scientific™ Replacement 96-well Clear Silicone Gaskets for Pierce™ ELIFA System
Tienda Thermo Scientific ProductosDescripción
These silicone gaskets are replacement accessories for the discontinued Thermo Scientific Pierce ELIFA System (Part No. 77000), which still may be used in various laboratories. The ELIFA (enzyme-linked immunoflow assay) system was a 96-well, vacuum-manifold apparatus for performing ELISA-type protocols on nitrocellulose membranes. The silicone gaskets are reusable accessories for this apparatus.
Related Products
Replacement Cannulas for Pierce™ ELIFA System
Related Products
Replacement Cannulas for Pierce™ ELIFA System
Especificaciones
Especificaciones
| Descripción | Replacement 96-well Clear Silicone Gaskets for Pierce™ ELIFA System, 4 Gaskets, The ELIFA System was a 96-well, Vacuum-manifold Apparatus for Performing ELISA-type Protocols on Nitrocellulose Membranes, Use with ELIFA System, Ideal for Protein Assays and Analysis |
| Para utilizar con (aplicación) | Protein biology, cell analysis, ELISA, protein assays and analysis, build your own immunoassay, ready-to-use immunoassay |
| Para utilizar con (equipo) | Sistema ELIFA |
| Cantidad | 4 Gaskets |
| Tipo | Replacement 96-well Clear Silicone Gaskets for Pierce™ ELIFA System |
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.