Descripción
Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.
Especificaciones
Especificaciones
| Antígeno | Renin |
| Aplicaciones | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.17 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | REN |
| N.º de referencia del gen | P00797, P06281 |
| Alias de gen | Angiotensinogenase, REN, Renin |
| Símbolos de los genes | REN, Ren1 |
| Mostrar más |