Learn More
STEMCELL Technologies ReLeSR™
Código de producto. p-7217803
Encompass_Preferred
Change view
Click to view available options
Cantidad:
500 mL
Tamaño de la unidad:
500 ml
15517014 500 mL 500 ml
1 options
Código de producto. 15517014
Marca: STEMCELL Technologies 05873
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Enzyme-free human ES and iPS cell selection and passaging reagent
  • ReLeSR™ is an enzyme-free reagent for dissociation and passaging of human embryonic stem (ES) or induced pluripotent stem (iPS) cells as aggregates without manual selection or scraping
  • Passaging human ES/iPS cells with ReLeSR™ easily generates optimally-sized aggregates, while eliminating the hassle and variability associated with manual manipulation
  • By eliminating the need for scraping, ReLeSR™ enables the use of culture flasks and other closed vessels, thus facilitating culture scale-up and automation
  • ReLeSR™, 500 mL, is now also manufactured following relevant cGMPs under a certified quality management system to ensure the highest quality and consistency for reproducible results
Especificaciones
Tipo de célula Pluripotent Stem Cells
Cantidad 500 mL