Learn More
Tuttnauer Release Trolley
Tienda Tuttnauer Productos
Click to view available options
:
Release Trolley; For use with: Tiva Washer
Descripción
- Suitable for the transport of washing trolley.
- Loading trolley with block system to load washing material into the washer disinfector.
- Anti-drop system to collect sinking water from load material.
- Wheels able to be blocked.
- Anti-upside-down system for security transport.
- Ergonomic handle and design.
- W x D x H: 634 x 633 x 880 mm (24.96 x 24.92 x 34.64 in.)
Especificaciones
Especificaciones
| Material | Acero inoxidable |
| Tipo de producto | Carro |
| Para utilizar con (equipo) | Tiva Washer |
| Cantidad | 1 Ea. |