Learn More
- Home
- Products
- Tubos
- Tubos de silicona y caucho
- Cole-Parmer Reinforced Thermoplastic Elastomer (TPE) Tubing
Cole-Parmer Reinforced Thermoplastic Elastomer (TPE) Tubing
Código de producto. 30054078 Tienda Cole-Parmer Productos
Change view
Click to view available options
Diámetro interior (imperial):
1/2 in
1/4 in
1/8 in
1 in
3/4 in
3/8 in
5/8 in
Diámetro exterior (imperial):
1-1/8 in
1-3/8 in
1/2 in
1 in
3/4 in
3/8 in
5/8 in
Longitud global (imperial):
15 ft
25 ft
50 ft
Tamaño de la unidad:
15 metros
4.50 metro
7.60 metro
19 product options available for selection
| Código de producto. | Diámetro interior (imperial) | Diámetro exterior (imperial) | Longitud global (imperial) | unitSize |
|---|---|---|---|---|
| 30054078 | 3/4 in | 1-1/8 in | 25 ft | 7.60 metro |
| 30054259 | 3/8 in | 5/8 in | 25 ft | 7.60 metro |
| 30053418 | 1/2 in | 3/4 in | 50 ft | 15 metros |
| 30053008 | 1/8 in | 3/8 in | 15 ft | 4.50 metro |
| 30054058 | 1 in | 1-3/8 in | 25 ft | 7.60 metro |
| 30053594 | 1/2 in | 3/4 in | 25 ft | 7.60 metro |
| 30053110 | 3/4 in | 1-1/8 in | 15 ft | 4.50 metro |
| 30053019 | 1/8 in | 3/8 in | 25 ft | 7.60 metro |
| 30054157 | 3/8 in | 5/8 in | 15 ft | 4.50 metro |
| 30054038 | 1/4 in | 1/2 in | 15 ft | 4.50 metro |
| 30053677 | 1/4 in | 1/2 in | 50 ft | 15 metros |
| 30053981 | 5/8 in | 1 in | 50 ft | 15 metros |
| 30053079 | 5/8 in | 1 in | 15 ft | 4.50 metro |
| 30053370 | 5/8 in | 1 in | 25 ft | 7.60 metro |
| 30054114 | 3/4 in | 1-1/8 in | 50 ft | 15 metros |
| 30053654 | 1/8 in | 3/8 in | 50 ft | 15 metros |
| 30053416 | 1/4 in | 1/2 in | 25 ft | 7.60 metro |
| 30053007 | 3/8 in | 5/8 in | 50 ft | 15 metros |
| 30053183 | 1 in | 1-3/8 in | 15 ft | 4.50 metro |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
Este artículo no se puede devolver.
Vea la política de devoluciones
Código de producto.
30054078
Proveedor
Cole-Parmer
N.º de proveedor
5011623
Inicie sesión o regístrese para comprobar su precio y disponibilidad.
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. E in., cellent for use in peristaltic pump applications. Great alternative to silicone tubing. Does not contain PVC, DEHP, phthalates, or other plasticizers. Ultra-low e in., tractables and leachables. Low gas and o in., ygen permeability. Low protein binding and nonpyrogenic material. Thermally weldable. USP Class VI, USP 661, and RoHS compliant and BPOG tested
Especificaciones
Especificaciones
| Certificaciones/conformidad | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diámetro interior (imperial) | 3/4 in |
| Material | TPE |
| Diámetro exterior (imperial) | 1-1/8 in |
| Longitud global (imperial) | 25 ft |
| Método de esterilización | 135°C |
| Color | Translúcido |