Learn More
Cole-Parmer Reinforced Thermoplastic Elastomer (TPE) Tubing
Código de producto. 30053110 Tienda Cole-Parmer Productos
Change view
Click to view available options
Diámetro interior (imperial):
1/2 in
1/4 in
1/8 in
1 in
3/4 in
3/8 in
5/8 in
Diámetro exterior (imperial):
1-1/8 in
1-3/8 in
1/2 in
1 in
3/4 in
3/8 in
5/8 in
Longitud global (imperial):
15 ft
25 ft
50 ft
Tamaño de la unidad:
15 metros
4.50 metro
7.60 metro
Código de producto. Diámetro interior (imperial) Diámetro exterior (imperial) Longitud global (imperial) unitSize
30053110 3/4 in 1-1/8 in 15 ft 4.50 metro
30054259 3/8 in 5/8 in 25 ft 7.60 metro
30053418 1/2 in 3/4 in 50 ft 15 metros
30053008 1/8 in 3/8 in 15 ft 4.50 metro
30054078 3/4 in 1-1/8 in 25 ft 7.60 metro
30054058 1 in 1-3/8 in 25 ft 7.60 metro
30053594 1/2 in 3/4 in 25 ft 7.60 metro
30053019 1/8 in 3/8 in 25 ft 7.60 metro
30054157 3/8 in 5/8 in 15 ft 4.50 metro
30054038 1/4 in 1/2 in 15 ft 4.50 metro
30053677 1/4 in 1/2 in 50 ft 15 metros
30053981 5/8 in 1 in 50 ft 15 metros
30053079 5/8 in 1 in 15 ft 4.50 metro
30053370 5/8 in 1 in 25 ft 7.60 metro
30054114 3/4 in 1-1/8 in 50 ft 15 metros
30053654 1/8 in 3/8 in 50 ft 15 metros
30053416 1/4 in 1/2 in 25 ft 7.60 metro
30053007 3/8 in 5/8 in 50 ft 15 metros
30053183 1 in 1-3/8 in 15 ft 4.50 metro
19 options
Código de producto. 30053110
Marca: Cole-Parmer 5011622
Este artículo requiere una actualización de precios antes de ser solicitado.
Solicite el precio de este artículo aquí.
Braided to withstand high-pressure applications
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. E in., cellent for use in peristaltic pump applications. Great alternative to silicone tubing. Does not contain PVC, DEHP, phthalates, or other plasticizers. Ultra-low e in., tractables and leachables. Low gas and o in., ygen permeability. Low protein binding and nonpyrogenic material. Thermally weldable. USP Class VI, USP 661, and RoHS compliant and BPOG tested
Especificaciones
Certificaciones/conformidad ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diámetro interior (imperial) 3/4 in
Material TPE
Diámetro exterior (imperial) 1-1/8 in
Longitud global (imperial) 15 ft
Método de esterilización EtO with no issues
Color Translúcido