Learn More
Cole-Parmer Reinforced Thermoplastic Elastomer (TPE) Tubing
Código de producto. 30053008 Tienda Cole-Parmer Productos
Change view
Click to view available options
Diámetro interior (imperial):
1/2 in
1/4 in
1/8 in
1 in
3/4 in
3/8 in
5/8 in
Diámetro exterior (imperial):
1-1/8 in
1-3/8 in
1/2 in
1 in
3/4 in
3/8 in
5/8 in
Longitud global (imperial):
15 ft
25 ft
50 ft
Tamaño de la unidad:
15 metros
4.50 metro
7.60 metro
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Diámetro interior (imperial) Diámetro exterior (imperial) Longitud global (imperial) unitSize
30053008 1/8 in 3/8 in 15 ft 4.50 metro
30054259 3/8 in 5/8 in 25 ft 7.60 metro
30053418 1/2 in 3/4 in 50 ft 15 metros
30054078 3/4 in 1-1/8 in 25 ft 7.60 metro
30054058 1 in 1-3/8 in 25 ft 7.60 metro
30053594 1/2 in 3/4 in 25 ft 7.60 metro
30053110 3/4 in 1-1/8 in 15 ft 4.50 metro
30053019 1/8 in 3/8 in 25 ft 7.60 metro
30054157 3/8 in 5/8 in 15 ft 4.50 metro
30054038 1/4 in 1/2 in 15 ft 4.50 metro
30053677 1/4 in 1/2 in 50 ft 15 metros
30053981 5/8 in 1 in 50 ft 15 metros
30053079 5/8 in 1 in 15 ft 4.50 metro
30053370 5/8 in 1 in 25 ft 7.60 metro
30054114 3/4 in 1-1/8 in 50 ft 15 metros
30053654 1/8 in 3/8 in 50 ft 15 metros
30053416 1/4 in 1/2 in 25 ft 7.60 metro
30053007 3/8 in 5/8 in 50 ft 15 metros
30053183 1 in 1-3/8 in 15 ft 4.50 metro
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Código de producto. 30053008 Proveedor Cole-Parmer N.º de proveedor 5011627
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Braided to withstand high-pressure applications
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. E in., cellent for use in peristaltic pump applications. Great alternative to silicone tubing. Does not contain PVC, DEHP, phthalates, or other plasticizers. Ultra-low e in., tractables and leachables. Low gas and o in., ygen permeability. Low protein binding and nonpyrogenic material. Thermally weldable. USP Class VI, USP 661, and RoHS compliant and BPOG tested
Especificaciones
Certificaciones/conformidad ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diámetro interior (imperial) 1/8 in
Material TPE
Diámetro exterior (imperial) 3/8 in
Longitud global (imperial) 15 ft
Método de esterilización 135°C
Color Translúcido