Learn More
Gilac™ Reinforced Finely Perforated Stackable Crate with Handles
Click to view available options
Dimensiones (Largo x Ancho x Alto):
545 x 390 x 200 mm
545 x 390 x 300 mm
Tamaño de la unidad:
cada uno
Descripción
- Stackable reinforced crate with handles, ideal for handling in the food industry
- Its very fine perforations ensure excellent ventilation of the food, even when stacked
- This professional-quality reinforced box is designed for intensive use
Especificaciones
Especificaciones
| Tipo de producto | Storage Container |
| Color | White |
| Material | HDPE |
| Forma | Rectangular |
| Dimensiones (Largo x Ancho x Alto) | 545 x 390 x 300 mm |
| Altura (métrico) | 300 mm |
| Longitud (métrico) | 545 mm |
| Anchura (métrico) | 390 mm |