missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ REG3B Polyclonal Antibody

Descripción
Reconstitute at 0.2 mg/mL in sterile PBS.
The REG3B gene encodes a protein that is part of the regenerating islet-derived (Reg) family, with significant roles in antimicrobial activity and tissue repair. REG3B is primarily expressed in the pancreas and is implicated in pancreatic regeneration, playing a critical role in conditions such as diabetes and pancreatitis. It exhibits potential therapeutic benefits in tissue injury and inflammatory disorders, including inflammatory bowel disease and acute liver injury. REG3B promotes acinar-to-ductal metaplasia through the activation of the RAS-RAF-MEK-ERK signaling pathway, suggesting its involvement in the early stages of pancreatic ductal adenocarcinoma development. Additionally, REG3B has been identified as an essential regulator of macrophage trafficking in cardiac healing post-myocardial ischemia, aiding in the recruitment and expansion of myeloid cells. Research aims to target REG3B and its receptors to interrupt pathological processes, offering promising avenues for treatment of pancreatic and extrapancreatic diseases.
Especificaciones
Especificaciones
| Antígeno | REG3B |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with trehalose and no preservative |
| génica | Reg3b |
| N.º de referencia del gen | P25031 |
| Alias de gen | HIP; Pancreatitis-associated protein 1; Pap; PAP1; peptide 23; Reg III-beta; Reg2; REG-2; Reg3b; REG-3-beta; regenerating islet-derived 3 beta; regenerating islet-derived protein 3 beta; regenerating islet-derived protein 3-beta; Regenerating islet-derived protein 3-beta 15 kDa form; Regenerating islet-derived protein 3-beta 16.5 kDa form; Regenerating islet-derived protein III-beta; REG-III; RegIII (beta) |
| Símbolos de los genes | Reg3b |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?