Learn More
Bellingham Stanley™ Eclipse Professional Grade Optical Hand-Held Refractometer
Descripción
- Range: antifreeze - % Ethylene Glycol 0 to 60; % Propylene Glocol 0 to 60
- Simple zero adjust with lock
- Shaded field of view for distinction
- High precision, clear scale
- Easy-clean prism
- Unique sample “dribble” feature
- Low weight
- Anti-roll supports
- Push-on prism flap
Especificaciones
Especificaciones
| Tipo | Refractómetros de mano |
| Scale Range | 0 to 60 Brix |
| Incrementos | 2.5 |
| Tipo de escala | Antifreeze - % concentration by vol., ethylene glycol and propylene glycol |
| Para utilizar con (aplicación) | Measurement of heat transfer fluids in air conditioning plants, heat exchangers, pasteurisers, and fluid based radiator systems, automobile manufacturer cooling system loading, and more |
| Incluye | Certificate of calibration, foam carry case and instruction manual |