missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This antibody is a rabbit polyclonal antibody raised against full length human REDD1 antigen. This antibody is specific to the REDD1 from siRNA experiment.
DDIT4 gene ontology annotations related to this gene include cellular response to dexamethasone stimulus; intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator; negative regulation of glycolytic process; negative regulation of TOR signaling; protein-containing complex disassembly; reactive oxygen species metabolic process; response to hypoxia.
Specifications
Specifications
| Antígeno | REDD1 |
| Aplicaciones | Western Blot, Immunoprecipitation |
| Clasificación | Polyclonal |
| Concentración | 0.53 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | DDIT4 |
| N.º de referencia del gen | Q8VHZ9, Q9D3F7, Q9NX09 |
| Alias de gen | DDIT4, Dig2, REDD 1, REDD1, RP11 442H21.1, RTP801 |
| Símbolos de los genes | DDIT4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?