missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Red Counterstain C

Código de producto. 30000775
Change view
Click to view available options
Cantidad:
50 mL
Tamaño de la unidad:
50 ml
Código de producto. Cantidad unitSize
30000775 50 mL 50 ml
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30000775

Marca: R&D Systems™ 48003019

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

R&D Systems™ Red Counterstain C is an acidophylic cytoplasmic counterstain that is included in the VasoTACS kit.

  • Provides an alternative red counterstain yielding a bright pink stain for vascular and smooth muscles tissues.
  • Hazard Statements (GHS-US):
    • H225 - Highly flammable liquid and vapor.
    • H319 - Causes serious eye irritation.
    • H371 - May cause damage to organs (optic nerve, central nervous system).
  • Precautionary Statements (GHS-US):
    • P210 - Keep away from extremely high or low temperatures, ignition sources, and incompatible materials. - No smoking
    • P233 - Keep container tightly closed.
    • P240 - Ground/bond container and receiving equipment
    • P241 - Use explosion-proof electrical, ventilating, and lighting equipment.
    • P242 - Use only non-sparking tools.
    • P243 - Take precautionary measures against static discharge.
    • P260 - Do not breathe vapors, mist, or spray.
    • P264 - Wash hands, forearms, and other exposed areas thoroughly after handling.
    • P270 - Do not eat, drink, or smoke when using this product.
    • P280 - Wear protective gloves, protective clothing, and eye protection.
    • P303+P361+P353 - IF ON SKIN (OR HAIR): Take off immediately all contaminated clothing. Rinse skin with water/shower.
    • P305+P351+P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P337+P313 - If eye irritation persists: Get medical advice/attention.
    • P370+P378 - In case of fire: Use appropriate media (see section 5) to extinguish.
    • P403+P235 - Store in a well-ventilated place. Keep cool.
    • P405 - Store locked up.
    • P501 - Dispose of contents/container in accordance with local, regional, national, territorial, provincial, and international regulations.
TRUSTED_SUSTAINABILITY

Especificaciones

Cantidad 50 mL
Contenido y almacenamiento Store the unopened product at room temperature
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.