Learn More
Gilac™ Rectangular HACCP Container with Lid
Click to view available options
Color:
Amarillo
Blanco
Rojo
Verde
Capacidad (métrico):
12 L
15 L
35 L
55 L
Tamaño de la unidad:
cada uno
Descripción
- HACCP rectangular food container.
- For large storage volumes, lid can support the weight of heavy loads.
- Multipurpose container.
- Rewritable identification cartridge for inventory tracking.
- No gaps nor overlaps.
- Machine washable.
- Recessed handles for better handling.
- Stackable without lid.
- Compatible with all GILAC 4 and 6 wheel dollies.
Especificaciones
Especificaciones
| Tipo | Storage Tote |
| Material | HDPE |
| Color | Amarillo |
| Forma | Rectangulares |
| Capacidad (imperial) | 4 gal. |
| Capacidad (métrico) | 15 L |