Descripción
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human REC8. This antibody can recognize the monomeric, non-phosphorylated form (63-70 kDa) of REC8 and other forms(80-100 kDa) which represent either post transla
This gene encodes a member of the kleisin family of SMC (structural maintece of chromosome) protein partners. The protein localizes to the axial elements of chromosomes during meiosis in both oocytes and spermatocytes. In the mouse, the homologous protein is a key component of the meiotic cohesion complex, which regulates sister chromatid cohesion and recombination between homologous chromosomes. Multiple alternatively spliced variants, encoding the same protein, have been found for this gene.
Especificaciones
Especificaciones
| Antígeno | REC8 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.21 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | REC8 |
| N.º de referencia del gen | O95072, Q8C5S7 |
| Alias de gen | Cohesin Rec8p, HR21spB, REC8, REC8 homolog (yeast), REC8L1, Rec8p |
| Símbolos de los genes | Rec8 |
| Mostrar más |