Learn More
DiscoverX Ready-to-Assay™ CCR4 Chemokine Receptor Frozen Cells
Descripción
The Ready-to-Assay™ CCR4 Chemokine Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. MIP, RANTES) based activation of CCR4 GPCR activity via detection of increasing levels of intracellular calcium. The included cells overexpress CCR4 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of CCR4 , the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the cells in a convenient thaw-and-use format.
Especificaciones
Especificaciones
| Entrada del ensayo | CCR4 |
| Para utilizar con (aplicación) | Frozen Cells |
| Agonista primario | MIP, RANTES |
| Cantidad | 2 viales de células |
| Entrada diana | CCR4 |