Learn More
Invitrogen™ REA Monoclonal Antibody (2B5)

Descripción
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human HEK293 whole cell, rat brain tissue, rat heart tissue, rat liver tissue, mouse brain tissue, mouse heart tissue, mouse liver tissue. IHC: human gallbladder adenocarcinoma tissue, human ovarian serous adenocarcinoma tissue, human renal clear cell carcinoma tissue, human lung cancer tissue, mouse colon tissue, rat colon tissue. Flow: HepG2 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Especificaciones
Especificaciones
| Antígeno | REA |
| Aplicaciones | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Monoclonal |
| Clon | 2B5 |
| Concentración | 500 μg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 4mg trehalose and no preservative |
| génica | PHB2 |
| N.º de referencia del gen | O35129, Q5XIH7, Q99623 |
| Alias de gen | AU044498; BAP; Bap37; Bap-37; Bcap27; BCAP37; B-cell associated protein; B-cell receptor associated protein 37; B-cell receptor-associated protein 37; B-cell receptor-associated protein BAP37; D-prohibitin; hBAP; MGC117268; p22; phb2; phb2 {ECO:0000250; PNAS-141; prohibitin 2; prohibitin-2; Rea; Repressor of estrogen receptor activity; UniProtKB:Q99623} |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.