Learn More
RDH10 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14644-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The RDH10 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RDH10. This antibody recognizes human, mouse, rat antigen. The RDH10 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Especificaciones
| RDH10 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 157506 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |