missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBPMS Antibody (OTI3B7), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-73836AF350
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
RBPMS Monoclonal specifically detects RBPMS in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Especificaciones
| RBPMS | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RBPMS | |
| Full length recombinant protein of human RBPMS (NP_001008712) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI3B7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| FLJ32971, Heart and RRM expressed sequence, Hermes, HERMESRNA-binding protein with multiple splicing, RBP-MS, RNA binding protein with multiple splicing | |
| Mouse | |
| Affinity Purified | |
| DNA Repair, Mismatch Repair | |
| 11030.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction