Learn More
Invitrogen™ RBPJ Monoclonal Antibody (6G4)
GREENER_CHOICE
Código de producto. 17941201
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
17941201 100 μg 100 microgramos
1 options
Código de producto. 17941201 Proveedor Invitrogen™ N.º de proveedor MA549168
Mouse Monoclonal Antibody
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HEK293 whole cell, human HEPG2 whole cell, human HELA whole cell, human MCF-7 whole cell, human U87 whole cell, human HL-60 whole cell, rat NRK whole cell, mouse smooth muscle tissue. Flow: HEPA1-6 cell, U937 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
CSL plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair. The protein is abundantly expressed in heart and skeletal muscle and expression is increased by passive stretch. Mutations in this gene can lead to X-linked deafness.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno RBPJ
Aplicaciones Flow Cytometry, Western Blot
Clasificación Monoclonal
Clon 6G4
Concentración 500 μg/mL
Conjugado Unconjugated
Formulación PBS with 4mg trehalose and no preservative
génica RBPJ
N.º de referencia del gen P31266, Q06330
Alias de gen AI843960; AOS3; CBF1; CBF-1; csl; H-2K binding factor-2; Igkjrb; Igkjrb1; Igkrsbp; immunoglobulin kappa J region recombination signal binding protein 1; J kappa-recombination signal-binding protein; KBF2; Rbpj; RBP-J; RBP-J kappa; RBPJK; RBP-JK; rbp-jkappa; RBPSUH; recombination signal binding protein for immunoglobulin kappa J region; recombining binding protein suppressor of hairless; recombining binding protein suppressor of hairless-like; recombining binding protein suppressor of hairless-like protein; renal carcinoma antigen NY-REN-30; SUH; suppressor of hairless homolog; Unknown (protein for MGC:128782)
Símbolos de los genes RBPJ
Especie del huésped Mouse
Inmunógeno E.coli-derived human RBPJK/RBPJ recombinant protein (Position: K41-Q467).
Método de purificación Antigen affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 19664, 3516, 679028
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG2b