missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This protein belongs to the lipocalin family and is the specific carrier for retinol (vitamin A alcohol) in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein posttranslationally and results in defective delivery and supply to the epidermal cells.
Specifications
Specifications
| Antígeno | RBP4 |
| Aplicaciones | Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 1D12B11 |
| Concentración | 1.21 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | RBP4 |
| N.º de referencia del gen | P02753 |
| Alias de gen | Plasma retinol binding protein, PRBP, RBP, RBP4, Retinol binding protein 4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?