Learn More
RBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16852045 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16852045 20 μL 20 microlitros
16842045 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16852045 Proveedor Proteintech N.º de proveedor 226831AP20UL
Rabbit Polyclonal Antibody
This gene encodes the carrier protein involved in the transport of retinol (vitamin A alcohol) from the liver storage site to peripheral tissue. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. Multiple transcript variants encoding different isoforms have been found for this gene.
Especificaciones
Antígeno RBP1
Aplicaciones Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.2 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica RBP1
N.º de referencia del gen P02696, P09455, Q00915
Alias de gen CRABP I, CRBP, CRBP I, CRBP1, CRBPI, RBP1, RBPC, Retinol binding protein 1
Símbolos de los genes Rbp1
Especie del huésped Rabbit
Inmunógeno RBP1 Fusion Protein Ag18572
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 19659, 25056, 5947
Especies diana Rat, Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG