Learn More
RBMS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16847883 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16847883 20 μL 20 microlitros
16837883 150 μL 150 microlitros
2 options
Código de producto. 16847883
Marca: Proteintech 110612AP20UL
Rabbit Polyclonal Antibody
This gene encodes a member of a small family of proteins which bind single stranded DNA/RNA. These proteins are characterized by the presence of two sets of ribonucleoprotein consensus sequence (RNP-CS) that contain conserved motifs, RNP1 and RNP2, originally described in RNA binding proteins, and required for DNA binding. These proteins have been implicated in such diverse functions as DNA replication, gene transcription, cell cycle progression and apoptosis. Several transcript variants, resulting from alternative splicing and encoding different isoforms, have been described. A pseudogene for this locus is found on chromosome 12.
Especificaciones
Antígeno RBMS1
Aplicaciones Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.25 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica RBMS1
N.º de referencia del gen P29558, Q5PQP1, Q91W59
Alias de gen C2orf12, HCC 4, MSSP, MSSP 1, MSSP 2, MSSP 3, MSSP1, RBMS1, SCR2, YC1
Símbolos de los genes Rbms1
Especie del huésped Rabbit
Inmunógeno RBMS1 Fusion Protein Ag1493
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 362138, 56878, 5937
Especies diana Rat, Mouse, Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG