Learn More
RBM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16852974 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16852974 20 μL 20 microlitros
16842974 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16852974 Proveedor Proteintech N.º de proveedor 143631AP20UL
Rabbit Polyclonal Antibody
This gene is a member of the glycine-rich RNA-binding protein family and encodes a protein with one RNA recognition motif domain. Expression of this gene is induced by cold shock and low oxygen tension. A pseudogene exists on chromosome 1. Multiple alternatively spliced transcript variants that are predicted to encode different isoforms have been characterized although some of these variants fit nonsense-mediated decay criteria.
Especificaciones
Antígeno RBM3
Aplicaciones Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.35 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica RBM3
N.º de referencia del gen O89086, P98179, Q925G0
Alias de gen IS1 RNPL, Putative RNA binding protein 3, RBM3, RNA binding motif protein 3, RNPL
Símbolos de los genes RBM3
Especie del huésped Rabbit
Inmunógeno RBM3 Fusion Protein Ag5934
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 114488, 19652, 5935
Especies diana Human, Rat, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG