Learn More
RBM17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16814534 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16814534 20 μL 20 microlitros
16804534 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16814534 Proveedor Proteintech N.º de proveedor 153741AP20UL
Rabbit Polyclonal Antibody
This gene encodes an RNA binding protein. The encoded protein is part of the spliceosome complex and functions in the second catalytic step of mRNA splicing. Alternatively spliced transcript variants have been described. Related pseudogenes exist on chromosomes 9 and 15.
Especificaciones
Antígeno RBM17
Aplicaciones Western Blot, Immunoprecipitation
Clasificación Polyclonal
Concentración 0.15 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica RBM17
N.º de referencia del gen Q8JZX4, Q96I25
Alias de gen 45 kDa splicing factor, RBM17, RNA binding motif protein 17, SPF45, Splicing factor 45
Símbolos de los genes Rbm17
Especie del huésped Rabbit
Inmunógeno RBM17 Fusion Protein Ag0985
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 291295, 76938, 84991
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG