missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBL2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27251-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Two retinoblastoma-related proteins, p107 and pRb2/p130 are structurally and functionally similar to the product of the retinoblastoma gene (pRb/p105). Like pRb they bind transforming proteins of DNA tumor viruses through a particular region called the "pocket domain". Both proteins play a fundamental role in growth control. The highest percentage of undetectable levels and the tightest inverse correlation with the histological grading and with PCNA expression in the most aggressive tumor types were found for pRb2/p130, which may suggest an important role for this protein in the pathogenesis and progression of lung cancer.Especificaciones
| RBL2 | |
| Polyclonal | |
| Unconjugated | |
| RBL2 | |
| P130, pRb2, Rb2, RBL2, RBR 2, retinoblastoma like 2 (p130), Retinoblastoma like protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5934 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q08999 | |
| RBL2 | |
| RBL2 Fusion Protein Ag25087 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido