Learn More
Invitrogen™ RBFOX2 Polyclonal Antibody
Descripción
RBFOX2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
This gene is one of several human genes similar to the C. elegans gene Fox-1. This gene encodes an RNA binding protein that is thought to be a key regulator of alternative exon splicing in the nervous system and other cell types. The protein binds to a conserved UGCAUG element found downstream of many alternatively spliced exons and promotes inclusion of the alternative exon in mature transcripts. The protein also interacts with the estrogen receptor 1 transcription factor and regulates estrogen receptor 1 transcriptional activity. Multiple transcript variants encoding different isoforms have been found for this gene.
Especificaciones
Especificaciones
| Antígeno | RBFOX2 |
| Aplicaciones | Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | Conc. Not Determined |
| Conjugado | Unconjugated |
| Formulación | PBS with 5mM sodium azide |
| génica | RBFOX2 |
| N.º de referencia del gen | A1A5R1, O43251, Q8BP71 |
| Alias de gen | dJ106I20.3; Fbm2; fox-1 homolog B; Fox-1 homolog Fxh; fox-1 homologue; FOX2; Fox-2; Fxh; Fyn-binding molecule 2; hexaribonucleotide-binding protein 2; HNRBP2; Hrnbp2; Rbfox2; Rbm9; repressor of tamoxifen transcriptional activity; RNA binding fox-1 homolog 2; RNA binding motif protein 9; RNA binding protein fox-1 homolog 2; RNA binding protein fox-1 homolog 2; RNA binding protein, fox-1 homolog (C. elegans) 2; RNA binding protein, fox-1 homolog (C. elegans) 2; RNA binding protein, fox-1 homolog 2; RNA-binding motif protein 9; RNA-binding protein 9; RTA; zgc:85694 |
| Símbolos de los genes | RBFOX2 |
| Mostrar más |