Learn More
RB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16886193 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16886193 20 μL 20 microlitros
16876193 150 μL 150 microlitros
2 options
Código de producto. 16886193
Marca: Proteintech 100482IG20UL
Rabbit Polyclonal Antibody
This antibody is a rabbit polyclonal antibody raised against human RB1 fusion protein.
Rb is a tumor suppressor nuclear phosphoprotein capable of binding to DNA. It is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus-16 E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth. Aberrations in the RB gene have been implicated in cancers of breast, colon, prostate, kidney, nasopharynx, and leukemia. Phosphorylation of Rb at S608 depends on CDK4.
Especificaciones
Antígeno RB1
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica RB1
N.º de referencia del gen P06400, P13405
Alias de gen OSRC, p105 Rb, pp110, pRb, RB, RB1, retinoblastoma 1
Símbolos de los genes RB1
Especie del huésped Rabbit
Inmunógeno RB1 Fusion Protein Ag0006
Método de purificación Protein A
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 19645, 5925
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG