Learn More
Invitrogen™ Ravulizumab Recombinant Monoclonal Antibody
Código de producto. 30283728
Change view
Click to view available options
Cantidad:
1 mg
100 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
30283728 1 mg 1 miligramo
30283396 100 μg 100 microgramos
2 options
Código de producto. 30283728
Marca: Invitrogen™ MA559286
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Ravulizumab is considered a long-acting complement 5 (C5) inhibitor that has been undergoing clinical trials for the treatment of paroxysmal nocturnal hemoglobinuria (PNH) as of 4 February, 2016. A pharmaceutical similar to ravulizumab (ALXN1210), called eculizumab, is currently approved for the treatment of PNH in 46 countries under the brand name Soliris®. Ravulizumab is considered by Alexion Pharmaceuticals Inc. to be a next-generation eculizumab molecule. Ravulizumab was subsequently approved by the US FDA in December of 2018 for a variety of beneficial characteristics that make it an advanced, next-generation agent in comparison to eculizumab. In particular, ravulizumab is currently the first and only long-acting C5 complement inhibitor that can be administered every eight weeks for the treatment of adult patients with PNH whereas eculizumab is a bi-weekly treatment, Label. Moreover, virtually all of the phase 3 trial results for ravulizumab have demonstrated the equivalent efficacy and safety established by eculizumab and that patients transition safely and effectively from using eculizumab to ravulizumab. Subsequently, whereas PNH patients may have needed to previously plan their lives rather strictly around the bi-weekly infusion administrations of eculizumab, with ravulizumab such patients can find a more relaxed dosing schedule of only six or seven infusions over an entire year, Label.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Ravulizumab Humanized
Aplicaciones ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Clasificación Recombinant Monoclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Alias de gen ALXN-1210; ravulizumab-cwvz
Especie del huésped Human
Método de purificación Protein A
Cantidad 1 mg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG2SA