Learn More
Invitrogen™ Rat NEFL ELISA Kit
Descripción
Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-L is the light or low molecular weight microfilament subunit and runs on SDS-PAGE gels at approximately 70 kDa. Neurofilament are the 10nm or intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. NF-H is the heavy or high molecular weight microfilament subunit and runs on SDS-PAGE gels in the range 180-220 kDa, with some variation in different species.
Especificaciones
Especificaciones
| Número de acceso | P19527 |
| Intervalo del ensayo | 15.63 to 1000 pg/mL |
| Sensibilidad del ensayo | 9.38 pg/mL |
| Conjugado | HRP |
| Método de detección | Colorimetric |
| Formato del kit de inmunoensayo | ELISA Microplate (Dismountable) |
| Tipo de producto | ELISA |
| Tipo de muestra | Plasma, Serum |
| Especies diana | Rat |
| Para utilizar con (equipo) | Colorimetric Microplate Reader |
| Mostrar más |