missing translation for 'onlineSavingsMsg'
Learn More

Rat Fas Ligand/TNFSF6 Antibody, R&D Systems™
GREENER_CHOICE

Código de producto. 18767987
Change view
Click to view available options
Cantidad:
25 μg
Tamaño de la unidad:
25 microgramos
Código de producto. Cantidad unitSize
18767987 25 μg 25 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18767987

Marca: R&D Systems AF1858SP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Goat Polyclonal Antibody

Fas Ligand/TNFSF6 Polyclonal specifically detects Fas Ligand/TNFSF6 in Rat samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Fas Ligand/TNFSF6
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N.º de referencia del gen P36940
Alias de gen apoptosis (APO-1) antigen ligand 1, Apoptosis antigen ligand, APT1LG1CD95L, APTL, CD178, CD178 antigen, CD95L, CD95-L, Fas antigen ligand, Fas ligand, Fas ligand (TNF superfamily, member 6), FASLCD95 ligand, FASLG, TNFSF6, TNFSF6FasL, tumor necrosis factor (ligand) superfamily, member 6, tumor necrosis factor ligand superfamily member 6
Símbolos de los genes FASLG
Especie del huésped Goat
Inmunógeno Mouse myeloma cell line NS0-derived recombinant rat Fas Ligand/TNFSF6 Leu104-Leu278 Accession # P36940
Método de purificación Affinity Purified
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 356
Especificidad de la prueba Detects rat Fas Ligand/TNFSF6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse Fas Ligand is observed and 5% cross-reactivity with recombinant human Fas Ligand is observed.
Reconstitución Reconstitute at 0.2 mg/mL in sterile PBS.
Especies diana Rat
Contenido y almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.