missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat CCR6 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Descripción
CCR6 Monoclonal specifically detects CCR6 in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antígeno | CCR6 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 876515R |
| Conjugado | Alexa Fluor 488 |
| Dilución | Flow Cytometry 0.25-1 μg/106 cells |
| Formulación | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| N.º de referencia del gen | Q5BK58 |
| Alias de gen | BN-1, CC-CKR-6, CCR6, CCR-6, CD196, CD196 antigen, chemokine (C-C motif) receptor 6, CKRL3, CKR-L3, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6, CMKBR6STRL22, DRY6, GPR29, GPR29C-C CKR-6, GPRCY4, GPR-CY4, GPRCY4DRY-6, lymphocyte, 22, STRL22 |
| Especie del huésped | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?