Learn More
CD209a Rat anti-Mouse, Brilliant Violet 650, Clone: 5H10, BD Optibuild™
Descripción
The 5H10 monoclonal antibody specifically binds to mouse CD209a, or CIRE, which is the most closely related homolog of human DC-SIGN. In the mouse, several homologs of human CD209 (DC-SIGN) have been described. CD209a is a 238-amino acid type-II transmembrane C-type lectin, with an extracellular carbohydrate recognition domain. It can bind to mannosylated ligands in a Ca++ dependent manner. CD209a is expressed by some conventional CD4+ and CD4-CD8- dendritic cells (DC) but not by CD8+ DC. It is also expressed on monocytes, splenic plasmacytoid pre-DC, macrophages, and resident peritoneal cells. CD209a expression is downregulated upon cellular activation. CD209a is thought to play a role in the internalization and presentation of antigens to initiate immune responses.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Especificaciones
Especificaciones
| Antígeno | CD209a |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 5H10 |
| Concentración | 0.2mg/mL |
| Conjugado | Brilliant Violet 650 |
| Descripción | CD209; Cire; DC-SIGN; Dcsign; DC-SIGN1; CDSIGN |
| Formulación | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Especie del huésped | Rat |
| Inmunógeno | Peptide Derived from Extracellular Region of CD209a/CIRE |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.