Learn More
CD103 Rat anti-Mouse, Brilliant Violet 605, Clone: M290, BD Optibuild™
Rat Monoclonal Antibody
Marca: BD Biosciences 740355
Descripción
The M290 antibody specifically binds to CD103, the α chain of α IEL β 7 integrin. CD103 has a unique and fairly restricted tissue distribution. It is expressed on almost all intestinal intraepithelial lymphocytes (IEL), dendritic epidermal T cells (DEC), subpopulations of peripheral T cells, and distinct subsets of fetal, neonatal, and adult thymocytes. E-cadherin is the epithelial cell ligand for α IEL β 7 integrin. The ordered expression of α IEL during thymocyte development (which occurs under the influence of the thymic epithelium), high level of α α IEL β 7-mediated adhesion in in vitro assays.
Especificaciones
| CD103 | |
| Monoclonal | |
| 0.2mg/mL | |
| Itgae; Integrin alpha-E; αE; alpha-E1; ITAE; Integrin αIEL chain; aM290 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| M290 | |
| Brilliant Violet 605 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse Intestinal Epithelial Cells | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only