missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASGRF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12958-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
RASGRF1 encodes a protein that is a guanine nucleotide exchange factor similar to the Saccharomyces cerevisiae CDC25 gene product. Functional analysis has demonstrated that this protein stimulates the dissociation of GDP from RAS protein. The studies of the similar gene in mouse suggested that the Ras-GEF activity of this protein in brain can be activated by Ca2+ influx, muscarinic receptors, and G protein beta-gamma subunit. Mouse studies also indicated that the Ras-GEF signaling pathway mediated by this protein may be important for long-term memory. Alternatively spliced transcript variants encoding distinct isoforms have been reported.Especificaciones
| RASGRF1 | |
| Polyclonal | |
| Unconjugated | |
| RASGRF1 | |
| CDC25, CDC25L, GNRP, GRF1, GRF55, H GRF55, PP13187, Ras GRF1, RASGRF1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 192213, 19417, 5923 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P27671, P28818, Q13972 | |
| RASGRF1 | |
| RASGRF1 Fusion Protein Ag4072 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido