missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: VVFSTERYNP ESLLQEGEGR LGKCSARVFF KNQKPRPTIN VTCTRLIEKK KRQQEDYLLY KQMKQLKNPL EIVSIPDNHG HIDPSLRLIW Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 78%.
This gene was identified as a retinoid acid (RA) receptor-responsive gene. It encodes a type 1 membrane protein. The expression of this gene is upregulated by tazarotene as well as by retinoic acid receptors. The expression of this gene is found to be downregulated in prostate cancer, which is caused by the methylation of its promoter and CpG island. Alternatively spliced transcript variant encoding distinct isoforms have been observed.
Especificaciones
Especificaciones
| Antígeno | RARRES1 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.3 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | RARRES1 |
| N.º de referencia del gen | P49788 |
| Alias de gen | 5430417P09Rik; AI662122; latexin-like; LXNL; PEIG1; PERG-1; phorbol ester-induced gene 1 protein; Rarres1; RAR-responsive protein TIG1; retinoic acid receptor responder (tazarotene induced) 1; retinoic acid receptor responder 1; retinoic acid receptor responder protein 1; Tazarotene-induced gene 1 protein; tazarotene-induced protein 1; TIG1 |
| Símbolos de los genes | RARRES1 |
| Mostrar más |
Título del producto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?
