missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RARG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11424-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Retinoic acid, a metabolite of vitamin A, is necessary for normal organogenesis but acts as a teratogen at high levels during embryonic and fetal development. Retinoic acid functions through its interaction with the nuclear protein, retinoic acid receptor (RAR). RAR belongs to the steroid and thyroid hormone superfamily of nuclear receptor proteins which exert their effects by binding to specific DNA response elements, thus regulating gene expression in target cells. RAR exists as three major subtypes: alpha, beta and gamma. Two main isoforms of RAR gamma exist: RAR gamma-1 has been found to be predomitly expressed in skin, while RAR gamma-2 has been found early in embryogenesis and in embryonal carcinoma cells. RAR gamma is also known as RARD. The corresponding gene for the retinoic acid receptor gamma 1 is NR1B3.Especificaciones
| RARG | |
| Polyclonal | |
| Unconjugated | |
| RARG | |
| NR1B3, RAR gamma, RARC, RARG, Retinoic acid receptor gamma, retinoic acid receptor, gamma | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19411, 5916, 685072 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P13631, P18911 | |
| RARG | |
| RARG Fusion Protein Ag1991 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido