missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAP2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5110269
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: MREYKVVVLG SGGVGKSALT VQFVTGSFIE KYDPTIEDFY RKEIEVDSSP SVLEILDTAG TEQFASMRDL YIKNGQGFIL VYSLVNQQSF QDIKPMRDQI IRVKRYERVP MILVGNKVDL EGEREVSYGE GKALAEEWSC PFMETSAKNK ASVDELFAEI VRQMNYAAQP NGDEGCCSAC VIL.
This intronless gene belongs to a family of RAS-related genes. The proteins encoded by these genes share approximately 50% amino acid identity with the classical RAS proteins and have numerous structural features in common. The most striking difference between the RAP and RAS proteins resides in their 61st amino acid: glutamine in RAS is replaced by threonine in RAP proteins. Evidence suggests that this protein may be polyisoprenylated and palmitoylated.
Especificaciones
| RAP2B | |
| Polyclonal | |
| Unconjugated | |
| RAP2B | |
| 4021402C18Rik; AA408554; Rap2b; RAP2B, member of RAS oncogene family; ras-related protein Rap-2b; small GTP binding protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 170923, 5912, 74012 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.97 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P61225, P61226, P61227 | |
| RAP2B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-183 of human RAP2B (NP_002877.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido