Learn More
Invitrogen™ RAP1A/RAP1B Recombinant Rabbit Monoclonal Antibody (24GB1030)
GREENER_CHOICE
Código de producto. 30275287
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
30275287 100 μL 100 microlitros
30275551 20 μL 20 microlitros
2 options
Código de producto. 30275287
Marca: Invitrogen™ MA557209
Rabbit Recombinant Monoclonal Antibody
RAP1A/RAP1B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno RAP1A/RAP1B
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 24GB1030
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica RAP1A
N.º de referencia del gen P61224, P62834, P62835, P62836, Q62636, Q99JI6
Alias de gen 2810443E11Rik; AI848598; C21KG; G-22K; GTP-binding protein smg p21A; GTP-binding protein smg p21B; GTP-binding protein SMG-P21A; K-REV; KREV1; Krev-1; OK/SW-cl.11; RAL1B; rap 1A; RAP1; RAP1A; RAP-1A; RAP1A, member of RAS oncogene family; Rap1b; RAP1B, member of RAS oncogene family; Ras family small GTP binding protein RAP1B; RAS related protein 1b; RAS-related protein 1a; Ras-related protein Krev-1; Ras-related protein Rap-1A; RAS-related protein RAP1B; ras-related protein Rap-1b; RAS-related protein-1a; small GTP binding protein; SMGP21
Símbolos de los genes RAP1A, RAP1B
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from human RAP1A/RAP1B (aa 100-150).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 109905, 171337, 215449, 295347, 5906, 5908
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG