missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAP1A Polyclonal Antibody

Descripción
It is expected to cross-react with zebrafish and C. elegans based on sequence homology of 100% and 94%, respectively. PA1-29451 has been successfully used in Western blot applications where a band of 25 kDa is detected. Positive control: rat brain tissue extract.
The product of this gene belongs to the family of RAS-related proteins. These proteins share approximately 50% amino acid identity with the classical RAS proteins and have numerous structural features in common. The most striking difference between RAP proteins and RAS proteins resides in their 61st amino acid: glutamine in RAS is replaced by threonine in RAP proteins. The product of this gene counteracts the mitogenic function of RAS because it can interact with RAS GAPs and RAF in a competitive manner. Two transcript variants encoding the same protein have been identified for this gene.
Especificaciones
Especificaciones
| Antígeno | RAP1A |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.95 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| génica | RAP1A |
| N.º de referencia del gen | P62834, P62835, P62836 |
| Alias de gen | AI848598; C21KG; G-22K; GTP-binding protein; GTP-binding protein smg p21A; GTP-binding protein SMG-P21A; KREV1; Krev-1; rap 1A; RAP1; RAP1A; RAP-1A; RAP1A, member of RAS oncogene family; RAS-related protein 1a; ras-related protein Krev-1; Ras-related protein Rap-1A; RAS-related protein-1a; SMGP21 |
| Símbolos de los genes | RAP1A |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?