missing translation for 'onlineSavingsMsg'
Learn More

PeproTech RANK (soluble) Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Código de producto. p-7245503
Change view
Click to view available options
Cantidad:
2 x 500 μg
25 μg
50 μg
500 μg
Tamaño de la unidad:
1 miligramo
25 microgramos
50 microgramos
500 microgramos
Código de producto. Cantidad unitSize
17844403 25 μg 25 microgramos
17804443 50 μg 50 microgramos
17834533 2 x 500 μg 1 miligramo
30054460 500 μg 500 microgramos
4 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17844403

Marca: PeproTech 500P144BT25UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MQIAPPCTSE KHYEHLGRCC NKCEPGKYMS SKCTTTSDSV CLPCGPDEYL DSWNEEDKCL LHKVCDTGKA LVAVVAGNST TPRRCACTAG YHWSQDCECC RRNTECAPGL GAQHPLQLNK DTVCKPCLAG YFSDAFSSTD KCRPWTNCTF LGKRVEHHGT EKSDAVCSSS LPARK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human sRANK Receptor. Anti-Human sRANK Receptor-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human sRANK Receptor by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human sRANK Receptor (500-P144) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human sRANK Receptor. Western Blot: To detect hsRANK Receptor by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hsRANK Receptor is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P144BT-1MG will be provided as 2 x 500 μg

RANK is a member of the tumor necrosis factor receptor (TNFR) family. The full length mouse RANK cDNA encodes a type I transmembrane protein with an extracellular and a cytoplasmic domain. The extracellular domain contains two potential N-linked glycosylation sites. RANK shares significant amino acid homology with other members of the TNFR family in its extracellular four cysteine-rich repeats. Human and murine RANK share 81% amino acid identity in their extracellular domains.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno RANK (soluble)
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.1-1.0 mg/mL
Conjugado Biotin
Formulación PBS with no preservative
génica TNFRSF11A
N.º de referencia del gen Q9Y6Q6
Alias de gen CD265; FEO; LOH18CR1; loss of heterozygosity, 18, chromosomal region 1; Ly109; mRANK; ODFR; OFE; OPTB7; Osteoclast differentiation factor receptor; OSTS; Paget disease of bone 2; PDB2; Rank; receptor activator of NF-kappaB; receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; RGD1563614; TNF receptor superfamily member 11a; Tnfrsf11a; TRANCER; TRANCE-R; tumor necrosis factor receptor superfamily member 11A; tumor necrosis factor receptor superfamily member 11a, NFKB activator; tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Símbolos de los genes TNFRSF11A
Especie del huésped Rabbit
Inmunógeno E.coli-derived, 19.3 kDa Recombinant Human sRANK Receptor.
Método de purificación Antigen affinity chromatography
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 8792
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.