missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAI3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA533798
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Percent identity with other species by BLAST analysis: Human (100%) Gorilla (94%) Gibbon (81%).
Retinoic acid plays a critical role in development, cellular growth, and differentiation. The specific function for this retinoic acid-induced gene has not yet been determined; however, it may play a role in embryonic development and epithelial cell differentiation. This gene is a member of the type 3 G protein-coupling receptor family, characterized by the signature 7-transmembrane domain motif. This gene may also be involved in interaction between retinoid acid and G protein signalling pathways. RAI3 has been reported in lung, kidney, colon, pancreas, prostate, and small intestine. ESTs have been isolated from amnion, breast, colon, eye, lung, nasal cavity, placenta, skin, uterus, and vessel libraries.
Especificaciones
| RAI3 | |
| Polyclonal | |
| Unconjugated | |
| GPRC5A | |
| GPRC5A, RAIG-1, Retinoic acid induced 3, Retinoic acid responsive, GPCR5A, RAI3, RAIG1, Retinoic acid-inducible gene 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9052 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q8NFJ5 | |
| GPRC5A | |
| Synthetic 16 amino acid peptide from N-terminal extracellular domain of human GPRC5A / RAI3. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido